missing translation for 'onlineSavingsMsg'
Learn More

Lithium Aluminum Hydride (Powder) 95.0+%, TCI America™

Catalog No. p-7131867
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L020325G 25 g
L0203100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L020325G Supplier TCI America Supplier No. L020325G

Chemical Identifiers

CAS 16853-85-3
Molecular Formula AlH4Li
Molecular Weight (g/mol) 37.95
MDL Number MFCD00011075
InChI Key OCZDCIYGECBNKL-UHFFFAOYSA-N
Synonym lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
PubChem CID 21226445
IUPAC Name lithium(1+) alumanuide
SMILES [Li+].[AlH4-]

Specifications

Color White
Melting Point 125°C
Quantity 25 g
Form Crystalline Powder
UN Number 1410
Formula Weight 37.95
Percent Purity ≥98.0% (T)
Chemical Name or Material Lithium Aluminum Hydride (Powder)
EINECSNumber (1)-0016
RTECSNumber BD0100000
TSCA Yes