missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium aluminum hydride, 1M solution in THF, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Chemical Name or Material | Lithium Aluminum hydride |
| Color | Gray |
| CAS | 16853-85-3, 109-99-9 |
| Molecular Formula | AlH4Li |
| Linear Formula | LiAlH4 |
| MDL Number | MFCD00011075 |
| Fieser | 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162 |
| Merck Index | 15, 344 |
| Density | 0.9000g/mL |
| Synonym | lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride |
| Show More |
RUO – Research Use Only