missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminium di-n-butylamide, 0.16M solution in 1,2-Dimethoxyethane, Thermo Scientific™

$766.46

Chemical Identifiers

CAS 15405-86-4
Molecular Formula C32H72AlLiN4
Molecular Weight (g/mol) 546.87
InChI Key ISBVOXPIPNIBOM-UHFFFAOYSA-N
Synonym lithium aluminum di-n-butylamide, lithium tetrakis dibutylamino alumanuide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane
PubChem CID 57369578
IUPAC Name lithium;tetrakis(dibutylamino)alumanuide
SMILES [Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC449191000
View Documents Promotion Details
N/A 100mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC449195000
View Documents Promotion Details
N/A 500mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

15405-86-4
546.87
lithium aluminum di-n-butylamide, lithium tetrakis dibutylamino alumanuide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane
lithium;tetrakis(dibutylamino)alumanuide
C32H72AlLiN4
ISBVOXPIPNIBOM-UHFFFAOYSA-N
57369578
[Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC

Specifications

15405-86-4
-6°C
ISBVOXPIPNIBOM-UHFFFAOYSA-N
lithium;tetrakis(dibutylamino)alumanuide
57369578
0.86
Lithium aluminium di-n-butylamide
C32H72AlLiN4
lithium aluminum di-n-butylamide, lithium tetrakis dibutylamino alumanuide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane
[Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC
546.87
546.87
0.8600g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
May damage fertility.
May damage the unborn child.
Harmful if inhaled.
May form explosive peroxides.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only