Learn More
Lithium aluminium di-n-butylamide, 0.16M solution in 1,2-Dimethoxyethane, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 449191000
| Quantity | 100mL |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 15405-86-4 | |
| 546.87 | |
| lithium aluminum di-n-butylamide, lithium tetrakis dibutylamino alumanuide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane | |
| lithium;tetrakis(dibutylamino)alumanuide |
| C32H72AlLiN4 | |
| ISBVOXPIPNIBOM-UHFFFAOYSA-N | |
| 57369578 | |
| [Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC |
Specifications
| Lithium aluminium di-n-butylamide | |
| 100mL | |
| -6°C | |
| ISBVOXPIPNIBOM-UHFFFAOYSA-N | |
| lithium;tetrakis(dibutylamino)alumanuide | |
| 57369578 | |
| Glass bottle | |
| 0.8600g/mL |
| 15405-86-4 | |
| C32H72AlLiN4 | |
| lithium aluminum di-n-butylamide, lithium tetrakis dibutylamino alumanuide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane | |
| [Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC | |
| 546.87 | |
| 546.87 | |
| 0.86 |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May damage fertility.
May damage the unborn child.
Harmful if inhaled.
May form explosive peroxides.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger