missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetylide ethylenediamine complex, 25 wt.% slurry in toluene, AcroSeal™

Catalog No. ac439881000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439881000 100 mL AcroSeal Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC439881000 Supplier Thermo Scientific Chemicals Supplier No. 439881000
Only null left

CAS: 6867-30-7 | C4H9LiN2 | 92.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6867-30-7 , 108-88-3
Molecular Formula C4H9LiN2
Molecular Weight (g/mol) 92.07
MDL Number MFCD00013183
InChI Key QJQWXTYPTBEPGS-UHFFFAOYSA-N
Synonym lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine
PubChem CID 2724010
IUPAC Name ethane-1,2-diamine; ethynyllithium
SMILES [Li]C#C.NCCN

Specifications

Boiling Point 111.0°C
Quantity 100 mL
Linear Formula HC≡CLi·H2NCH2CH2NH2
Flash Point −14°C
Formula Weight 92.07
Packaging AcroSeal Glass Bottle
Chemical Name or Material Lithium acetylide ethylenediamine complex
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated ex
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 229-967-9

RUO – Research Use Only