missing translation for 'onlineSavingsMsg'
Learn More

Lithium AA Standard, 1000 ppm Li in 3% HCl, Ricca Chemical

$112.43 - $112.43

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium(1+) carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
ALI1KH-100
View Documents
Ricca Chemical Company
ALI1KH100
100 mL Natural LDPE Bottle
Each for $112.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

554-13-2
73.89
XGZVUEUWXADBQD-UHFFFAOYSA-L
CHEBI:6504
[Li+].[Li+].[O-]C([O-])=O
CLi2O3
MFCD00011084
11125
dilithium(1+) carbonate

Specifications

Natural LDPE Bottle
1,000ppm Li
97.0,4.79,0.52
Liquid
MFCD00011084
Approximately 0°C
[Li+].[Li+].[O-]C([O-])=O
73.89
11125
Lithium AA Standard
554-13-2,7647-01-0,7732-18-5
93.0,4.6,0.49
1 mL = 1 mg Li
CLi2O3
Colorless
XGZVUEUWXADBQD-UHFFFAOYSA-L
dilithium(1+) carbonate
100 mL
CHEBI:6504
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'