missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium 6, 6Li, plasma standard solution, Specpure™, 6Li 100μg/mL, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| UN Number | UN3264 |
| Quantity | 100 mL |
| Grade | Specpure |
| Chemical Name or Material | Lithium 6, 6Li, plasma standard solution, 6Li 100μg/mL |
Safety and Handling
| Hazard Category | H290-H315-H319 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390 |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only