missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lissamine rhodamine B sulfonyl chloride, pure

Catalog No. p-153852
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413230010 1 g Glass bottle
AC413230050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413230010 Supplier Thermo Scientific Chemicals Supplier No. 413230010
Only null left

CAS: 62796-29-6 | C27H29ClN2O6S2 | 577.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62796-29-6
Molecular Formula C27H29ClN2O6S2
Molecular Weight (g/mol) 577.107
MDL Number MFCD00042001
InChI Key YERWMQJEYUIJBO-UHFFFAOYSA-N
Synonym Sulforhodamine B sulfonyl chloride
PubChem CID 65223
IUPAC Name 5-chlorosulfonyl-2-[3-(diethylamino)-6-diethylazaniumylidenexanthen-9-yl]benzenesulfonate
SMILES CCN(CC)C1=CC2=C(C=C1)C(=C3C=CC(=[N+](CC)CC)C=C3O2)C4=C(C=C(C=C4)S(=O)(=O)Cl)S(=O)(=O)[O-]

Specifications

Assay Percent Range 99+%
Infrared Spectrum Authentic
Formula Weight 577.11
Grade Pure
Packaging Glass bottle
UV Spectrometry Molar ext.%gt; 85, 000L/mol.cm(lambda 564 +/- 3nm) in dichloromethane
Color Green
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Lissamine rhodamine B sulfonyl chloride, 99+%
Hazard Category PRODUCT NEEDS INERT ATMOSPHERE
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 263-735-8
TSCA TSCA

RUO – Research Use Only