accessibility menu, dialog, popup

UserName

CH3NCH3OSOOOSClOONH3CH3C(?)

CAS RN 62796-29-6

IUPAC Name: 9-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium
Synonyms: 9-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium
Molecular Weight (g/mol): 577.11
Molecular Formula: C27H29ClN2O6S2
InChi Key: YERWMQJEYUIJBO-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S(Cl)(=O)=O)=C2C=C1)N(CC)CC
Color 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)