missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lisinopril Dihydrate 98.0+%, TCI America™
$177.08 - $529.40
Chemical Identifiers
| Molecular Formula | C21H33N3O6 |
|---|---|
| Molecular Weight (g/mol) | 423.51 |
| MDL Number | MFCD01698825 |
| InChI Key | BAVDEDVBIHTHJQ-UVJOBNTFSA-N |
| Synonym | 1-[Nalpha-[(S)-1-Carboxy-3-phenylpropyl]-L-lysyl]-L-proline |
| PubChem CID | 22887897 |
| IUPAC Name | (2S)-1-[(2S)-6-amino-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]hexanoyl]pyrrolidine-2-carboxylic acid;hydrate |
| SMILES | C1CC(N(C1)C(=O)C(CCCCN)NC(CCC2=CC=CC=C2)C(=O)O)C(=O)O.O |
Chemical Identifiers
| C21H33N3O6 | |
| MFCD01698825 | |
| 1-[Nalpha-[(S)-1-Carboxy-3-phenylpropyl]-L-lysyl]-L-proline | |
| (2S)-1-[(2S)-6-amino-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]hexanoyl]pyrrolidine-2-carboxylic acid;hydrate |
| 423.51 | |
| BAVDEDVBIHTHJQ-UVJOBNTFSA-N | |
| 22887897 | |
| C1CC(N(C1)C(=O)C(CCCCN)NC(CCC2=CC=CC=C2)C(=O)O)C(=O)O.O |
Specifications
| Powder/Crystals | |
| MFCD01698825 | |
| 148°C | |
| BAVDEDVBIHTHJQ-UVJOBNTFSA-N | |
| (2S)-1-[(2S)-6-amino-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]hexanoyl]pyrrolidine-2-carboxylic acid;hydrate | |
| 1 g | |
| 405.50 | |
| Lisinopril Dihydrate |
| C21H33N3O6 | |
| White/Yellow | |
| 1-[Nalpha-[(S)-1-Carboxy-3-phenylpropyl]-L-lysyl]-L-proline | |
| C1CC(N(C1)C(=O)C(CCCCN)NC(CCC2=CC=CC=C2)C(=O)O)C(=O)O.O | |
| 423.51 | |
| 22887897 | |
| ≥98.0% (T) |
Safety and Handling
RTECSNumber : TW3589990
TSCA : No
Recommended Storage : Refrigerator