missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIS1 Recombinant Rabbit Monoclonal Antibody (23GB2825)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553527
Description
LIS1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
VAMP7 is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family, localizing to late endosomes and lysosomes. VAMP7 is thought to mediate the fusion of endosomes to their target lysosomes as well as other exocytosis events during phagocytosis and neuritogenesis. VAMP7 interacts with the VPS9 ankyrin repeat protein VARP, a protein that localizes to early endosomes and thought to regulate endosome dynamics. Together with CD82, VAMP7 can modulate the signaling of EGFR by regulating its endocytosis from the plasma membrane.Chemical Identifiers
| LIS1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| PAFAH1B1 | |
| HAMAP-Rule:MF_03141}; Lis1; Lis-1; LIS-1 {ECO:0000255; LIS2; lissencephaly 1 protein; lissencephaly-1 protein; lissencephaly-1 protein {ECO:0000255; MDCR; MDS; Mdsh; MGC25297; Miller-Dieker syndrome chromosome region; MMS10-U; Ms10u; NudF; PAF acetylhydrolase 45 kDa subunit; PAF acetylhydrolase 45 kDa subunit {ECO:0000255; PAFAH; PAF-AH 45 kDa subunit; PAF-AH 45 kDa subunit {ECO:0000255; PAFAH alpha; PAF-AH alpha; PAFAH alpha {ECO:0000255; PAF-AH alpha {ECO:0000255; PAF-AH beta; Pafah1b1; Pafaha; platelet activating factor acetylhydrolase 1b regulatory subunit 1; platelet activating factor acetylhydrolase 45 kDa subunit brain isoform; platelet activating factor acetylhydrolase 45 kDa subunit brain isoform variant; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa); platelet-activating factor acetylhydrolase beta subunit (PAF-AH beta); platelet-activating factor acetylhydrolase IB subunit alpha; platelet-activating factor acetylhydrolase IB subunit alpha {ECO:0000255; platelet-activating factor acetylhydrolase, isoform 1b, beta1 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 1; platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD); platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit 45kDa; platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa); RP23-194P5.2 | |
| PAFAH1B1 | |
| Primary | |
| Affinity chromatography |
| 23GB2825 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P43034, P63004, P63005 | |
| 18472, 5048, 83572 | |
| A synthesized peptide derived from human LIS1 (55-90AA). | |
| Antibody |
Specifications
| LIS1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PAFAH1B1 | |
| HAMAP-Rule:MF_03141}; Lis1; Lis-1; LIS-1 {ECO:0000255; LIS2; lissencephaly 1 protein; lissencephaly-1 protein; lissencephaly-1 protein {ECO:0000255; MDCR; MDS; Mdsh; MGC25297; Miller-Dieker syndrome chromosome region; MMS10-U; Ms10u; NudF; PAF acetylhydrolase 45 kDa subunit; PAF acetylhydrolase 45 kDa subunit {ECO:0000255; PAFAH; PAF-AH 45 kDa subunit; PAF-AH 45 kDa subunit {ECO:0000255; PAFAH alpha; PAF-AH alpha; PAFAH alpha {ECO:0000255; PAF-AH alpha {ECO:0000255; PAF-AH beta; Pafah1b1; Pafaha; platelet activating factor acetylhydrolase 1b regulatory subunit 1; platelet activating factor acetylhydrolase 45 kDa subunit brain isoform; platelet activating factor acetylhydrolase 45 kDa subunit brain isoform variant; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa); platelet-activating factor acetylhydrolase beta subunit (PAF-AH beta); platelet-activating factor acetylhydrolase IB subunit alpha; platelet-activating factor acetylhydrolase IB subunit alpha {ECO:0000255; platelet-activating factor acetylhydrolase, isoform 1b, beta1 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 1; platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD); platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit 45kDa; platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa); RP23-194P5.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18472, 5048, 83572 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2825 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P43034, P63004, P63005 | |
| PAFAH1B1 | |
| A synthesized peptide derived from human LIS1 (55-90AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |