Learn More
Agilent BioTek Liquid Handling Control Software
Catalog No. p-200007355
Change view
Click to view available options
Includes:
E-delivery, 5 Installation Licenses
E-delivery, 5 Installation Licenses, 21CFR Part 11 Compliance
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
BTLHC3 E-delivery, 5 Installation Licenses
BTLHC3SECUR E-delivery, 5 Installation Licenses, 21CFR Part 11 Compliance
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTLHC3 Supplier Agilent BioTek Supplier No. LHC3
Only null left
Liquid Handling Control Software provides a powerful yet flexible interface for use with washers, dispensers and combination liquid handling systems.
  • LHC software makes it easy to program, even the most complex protocols with a choice of deployment
  • Protocols can be run directly from the computer or downloaded to the instrument for convenient onboard recall and operation
  • Full instrument control
  • For use with 405 TS Washer, 405 LS Washer, ELx405 Select Deep Well Washer, 50 TS Washer, MultiFlo Dispenser, MultiFlo FX Dispenser, EL406 Washer Dispenser, 406 FX Washer Dispenser
  • Batch processing with connected BioStack Microplate Stacker or BioSpa 8 Automated Incubator
  • Seamless robotic integration to commercially available robotic systems
Specifications
Includes E-delivery, 5 Installation Licenses
Type Liquid Handling Control Software
For Use With (Equipment) Agilent BioTek Microplate Washers, Dispensers, and Liquid Handling Systems