missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Lipopolysaccharide (LPS) Solution (500X)

Catalog No. 501129325
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501129325 4 x 100 μL
501122025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501129325 Supplier Invitrogen™ Supplier No. 00497603

Intended for in vitro culture of B cells, monocytes, macrophages, and other cells that are responsive to LPS.

  • A ready to use solution of LPS from Escherichia coli 026:B6 in aqueous solution
  • LPS is a major component of the cell wall of gram negative bacteria
  • It is highly immunogenic and strongly activates immune cells bearing the CD14/TLR4/MD2 receptor complex
  • This reagent is intended for use in in vitro culture of B cells, monocytes, macrophages and other cells that are responsive to LPS

Specifications

Cell Type B cells, monocytes, macrophages and other cells that are responsive to LPS.
Concentration 500 X
Form Liquid
Product Type LPS Solution
Sterility Sterile
Product Line eBioscience
Quantity 4 x 100 μL

For Research Use Only. Not for use in diagnostic procedures.