missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ Stem Transfection Reagent

Catalog No. STEM00015
Encompass
Change view
Click to view available options
Quantity:
0.1 mL
0.3 mL
0.75 mL
1.5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STEM00015 1.5 mL
STEM00003 0.3 mL
STEM00008 0.75 mL
STEM00001 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. STEM00015 Supplier Invitrogen™ Supplier No. STEM00015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lipofectamine Stem Transfection Reagent is optimized to achieve maximum efficiency with minimal early differentiation in a wide range of stem cells. It can co-deliver DNA, RNA, and Cas9 ribonucleoprotein (RNP) complexes.

Lipofectamine Stem Transfection Reagent is optimized to achieve maximum efficiency with minimal early differentiation in a wide range of stem cells. It can co-deliver DNA, RNA, and Cas9 ribonucleoprotein (RNP) complexes. Lipofectamine Stem reagent is compatible with a variety of media systems, including feeder-free, helping support and simplify your stem cell culture workflow. Researchers can achieve up to 80% or better transfection efficiency in pluripotent stem cells (PSCs) and neural stem cells (NSCs), and up to 45% in mesenchymal stem cells (MSCs).

Transfecting stem cells without inhibiting cell viability and cell growth can be challenging due to the sensitivity of these cells. Lipofectamine Stem is a low toxicity reagent, requiring low amounts of nucleic acid, allowing your stem cells to stay healthy and continue proliferating without inducing differentiation. Lipofectamine Stem reagent is:

  • Superior in performance—up to 3-fold higher efficiency across iPSCs, hESCs, NSCs, and MSCs compared to other reagents
  • Versatile—one reagent for DNA, RNA, Cas9 protein, and co-transfection
  • Gentle on cells—maintains cell proliferation without inducing differentiation
  • Flexible—transfects cells in adherence or suspension—an alternative to electroporation

Achieve high efficiency gene editing in stem cells

Gene editing applications typically require the ability to co-deliver DNA, RNA, and/or protein into cells. Lipofectamine Stem Transfection Reagent is designed to co-deliver CRISPR/Cas9 complexes into human stem cells for high efficiency gene editing. This one reagent can be used to transfect large DNA plasmids such as all-in-one CRISPR/Cas9 vectors and co-deliver a range of RNAs, such as large Cas9 mRNA with tracr/crRNA complexes or single guide (sg) RNA, and protein such as Cas9 protein combined with tracr/crRNA or sgRNA.

A low-toxicity alternative to electroporation

Transfecting stem cells via electroporation methods can be challenging and cumbersome. Electroporation protocols recommend single cell suspension culture to ensure maximum efficiency and minimal toxicity. Using the reverse transfection protocol, the Lipofectamine Stem Transfection Reagent proves to be an effective and less toxic alternative to electroporation for stem cells. It requires low amounts of nucleic acid to achieve maximum transfection efficiency, on par with electroporation, while supporting continued proliferation without inducing differentiation.

Specifications

Classification Animal Origin-Free
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light
Cell Type Stem Cells
Sample Type Cells
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
Product Line Lipofectamine
Product Type Transfection Reagent
Quantity 1.5 mL
Serum Compatible No
Shipping Condition Wet Ice
Show More Show Less
With Lipofectamine Stem Transfection Reagent, how can I improve the transfection efficiency in my stem cells?

To obtain the highest percent of stem cells transfected with Lipofectamine Stem Transfection Reagent, we highly recommend that you follow cell culture conditions that promote formation of a monolayer versus clumps of cells. If the density of your stem cells is greater than 50% or they are being grown as colonies in other media, more Lipofectamine Stem Transfection Reagent may improve transfection efficiency. For additional details, please refer to the Lipofectamine Stem Transfection Reagent protocol pertinent to your cell type and media on the product page (https://www.thermofisher.com/order/catalog/product/STEM00008?ICID=search-product), under Documents, Product Literature.

Can I co-transfect plasmid and Cas9 protein (RNP) with Lipofectamine Stem Transfection Reagent?

Yes. Lipofectamine Stem Transfection Reagent can be used to co-deliver multiple payloads, including DNA, mRNA, and Cas9 protein (RNP) into stem cells.

At what density should I plate my stem cells before transfecting them using Lipofectamine Stem Transfection Reagent?

Cell confluency can impact the amount of Lipofectamine Stem Transfection Reagent that is needed for efficient transfection. Stem cells that are near confluency do not transfect well. Plating density should be such that cells are not overgrown 24 hours after transfection. To obtain the highest percent of stem cells transfected, we highly recommend that you dissociate both colony type and monolayer cultures into small clumps of 3-5 cells. If you are growing human pluripotent stem cells in a 24-well plate in Essential 8 Medium on Vitronectin, plate the cells to achieve 30-60% confluency on the day of transfection. For additional details, please refer to the Lipofectamine Stem Transfection Reagent protocol pertinent to your cell type and media on the product page (https://www.thermofisher.com/order/catalog/product/STEM00008?ICID=search-product), under Documents, Product Literature.

Can I use Lipofectamine Stem Transfection Reagent to transfect neural stem cells (NSCs)?

Yes. Lipofectamine Stem Transfection Reagent can be used to transfect a wide range of stem cell types, while supporting continued proliferation without inducing differentiation. It is our recommended solution for transfecting neural stem cells.

I used Lipofectamine Stem Transfection Reagent. Why are only the outer cells of my stem cell colonies transfected ?

For cells that have aggregated into large colonies, transfection attempts may result in ONLY the outer cells of the colonies being successfully transfected. Transfection in suspension at the time of passaging and re-plating can maximize access of the reagent to all the cells and increase transfection efficiency. We recommend cell culture conditions that promote formation of a monolayer versus clumps of cells. To avoid cell aggregates that form large colonies, gently dissociate colonies to small clumps of 3-5 cells to promote efficient transfection. Follow recommended seeding densities for the cell culture medium and passaging reagents used so that cells are at the recommended starting confluency of 30-60% at the time of transfection and have room to grow for 24-48 hours during transfection.

Can I use Lipofectamine Stem Transfection Reagent to transfect large plasmids?

Yes. Lipofectamine Stem Transfection Reagent can deliver small and large (up to 11 kb) DNA plasmids into stem cells with very low cytotoxicity.

Which serum-free medium should I use when I prepare Lipofectamine Stem Transfection Reagent-DNA complexes?

Certain components of culture media can be inhibitory to lipid-based transfections. For best results, we recommend using OptiMEM Reduced Serum Medium for preparing Lipofectamine Stem Transfection Reagent -DNA, -mRNA, and -Cas9 RNP transfection complexes.

Can I use Lipofectamine Stem Transfection Reagent for genome editing?

Yes. Lipofectamine Stem Transfection Reagent is optimized for genome editing in human pluripotent stem cells and neural stem cells. Lipofectamine Stem Transfection Reagent provides efficient transfection of large DNA plasmids such as CRISPR/Cas9 vectors, RNA such as Cas9 mRNA co-transfected with tracr/guide RNA, or Ribonucleoprotein complexes of Cas9 protein with guide RNAs.

How should I prepare my stem cells when I use Lipofectamine Stem Transfection Reagent?

Proliferating stem cells in culture need room to expand. We recommended plating cells so they will be 30-60% confluent at the time of transfection. For additional details, please refer to the Lipofectamine Stem Transfection Reagent protocol pertinent to your cell type and media on the product page (https://www.thermofisher.com/order/catalog/product/STEM00008?ICID=search-product), under Documents, Product Literature.

Can I use the same amount of DNA and Lipofectamine Stem Transfection Reagent for transfecting all stem cells?

When planning your transfection-based experiments, it is important to optimize the amount of nucleic acid and transfection reagent for each stem cell type being used, so that you may balance optimum transfection efficiency with minimal cytotoxicity.

Can Lipofectamine Stem Transfection Reagent be used for siRNA delivery?

We recommend using Lipofectamine RNAiMAX Transfection Reagent for siRNA delivery.

What media can I use with Lipofectamine Stem Transfection Reagent?

We recommend using our feeder-free Essential 8 Medium or StemFlex Medium on Vitronectin, Geltrex or Laminin-521 matrices. Lipofectamine Stem Transfection Reagent also supports other media systems such as mTeSR1 on Geltrex matrix.

Is Lipofectamine Stem Transfection Reagent compatible with feeder-free media, such as StemFlex Medium?

Yes. Lipofectamine Stem Transfection Reagent has been tested with StemFlex Medium and Essential 8 Medium on Vitronectin, Geltrex, and Laminin-521 matrices.

Can Lipofectamine Stem Transfection Reagent be used to transfect stem cells in suspension?

Yes. High transfection efficiency can be achieved in stem cells in suspension with reverse transfection using Lipofectamine Stem Transfection Reagent. Stem cells can easily be transfected at the time of passaging and during re-plating, when they are in suspension. For simple instructions on using reverse transfection with Lipofectamine Stem Transfection Reagent, please take a look at the transfection protocols for pluripotent stem cells, listed under Documents, Product Literature on the product page (https://www.thermofisher.com/order/catalog/product/STEM00008?ICID=search-product).

Can I use serum during transfection with Lipofectamine Stem Transfection Reagent?

Yes. Most albumin- and serum-supplemnted media do not inhibit transfection with this reagent.

What types of molecules can be transfected using Lipofectamine Stem Transfection Reagent?

Lipofectamine Stem Transfection Reagent can be used to deliver and co-deliver DNA, mRNA, and Cas9 protein (RNP) into stem cells, including neural stem cells (NSCs) and mesenchymal stem cells (MSCs).

What is the difference between Lipofectamine Stem Transfection Reagent and Lipofectamine 3000 Transfection Reagent?

Lipofectamine Stem Transfection Reagent is a lipid-based transfection reagent that is specifically designed for stem cells. It is our most versatile, and preferred choice for delivering DNA, mRNA, and Cas9 protein (RNP) into human pluripotent stem cells, as well as neural stem cells (NSCs) and mesenchymal stem cells (MSCs). Lipofectamine 3000 Transfection Reagent can be used to deliver DNA or RNA and is recommended for hard-to-transfect cell lines and workhorse cell lines.


For Research Use Only. Not for use in diagnostic procedures.