missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent

Catalog No. cmax00008
Change view
Click to view available options
Quantity:
0.1 mL
0.75 mL
1.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CMAX00008 0.75 mL
CMAX00001 0.1 mL
CMAX00015 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CMAX00008 Supplier Invitrogen™ Supplier No. CMAX00008
Only null left

Lipofectamine CRISPRMAX Cas9 Transfection Reagent is an optimized lipid-based transfection reagent for CRISPR-Cas9 protein delivery, providing the cleavage efficiency of electroporation with simplicity and scalability.

Lipofectamine CRISPRMAX Cas9 Transfection Reagent is the first optimized lipid-based transfection reagent for CRISPR knockout applications. It effectively delivers the Cas9 protein and guide RNA (gRNA) ribonucleoprotein (RNP) complex, ensuring high transfection efficiency and effective gene knockout. The proprietary lipid-based transfection reagent minimizes cellular toxicity, making it suitable to for sensitive cells. Its ease of use and compatibility with high-throughput screening make it a cost-effective alternative to electroporation.

Combined with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA, it provides:

  • Optimized for CRISPR—specially formulated for CRISPR-Cas9 RNP applications
  • Superior gene knockout—efficient Cas9 transfection ensures higher genomic cleavage
  • Broad cell type applicability—versatile for use in a wide range of cell lines, including primary cells and stem cells
  • Minimal cytotoxicity—viable, healthy cells maintained throughout Cas9 transfection
  • High-throughput compatible—an ideal delivery solution for functional genomic studies with CRISPR-Cas9

Achieve high transfection efficiency in novel genome editing applications

Lipofectamine CRISPRMAX Transfection Reagent enables successful Cas9 transfection in a wide range of cell types, including iPSC, mESC, N2A, CHO, A549, HCT116, HeLa, HEK293, and more. Unlike CRISPR plasmid or Cas9 mRNA, this protein-based approach eliminates the need for transcription or translation, reducing the risk of genomic integration and making the process cell cycle-independent. When combined with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA, it ensures superior cleavage efficiency and effective gene editing.

Specifications

Classification Animal Origin-Free
Content And Storage Contains one vial of Lipofectamine CRISPRMAX Reagent and one vial of Cas9 Plus Reagent.

Store at 2–8°C. Do not freeze.
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Protein, Ribonucleoprotein, Cas9 Ribonucleoprotein, Cas9, Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
Product Line CRISPRMAX, Lipofectamine
Product Type Transfection Reagent
Quantity 0.75 mL
Serum Compatible Yes
Shipping Condition Approved for shipment on Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.