missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liothyronine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 6893-02-3 |
| Linear Formula | C15H12I3 NO4 |
| MDL Number | MFCD00002593 |
| Melting Point | 234°C to 238°C |
| Synonym | 3,3 ′,5-Triiodo-L-thyronine; O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine; Liothyronine; T3 |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Formula Weight | 650.97 |
| Grade | Certified Reference Material |