missing translation for 'onlineSavingsMsg'
Learn More
Learn More
γ-Linolenic acid, ≥98.5% (GC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 3132-99-8 |
| Flash Point | 110°C - closed cup |
| Shipping Condition | Ambient |
| Physical Form | Neat |
| Linear Formula | C18H30O2 |
| MDL Number | MFCD00065718 |
| Synonym | cis,cis,cis-6,9,12-Octadecatrienoic acid |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 278.43 |
| Show More |