missing translation for 'onlineSavingsMsg'
Learn More

γ-Linolenic acid, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014367
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014367 100 mg
111014368 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014367 Supplier Merck Emd Millipore Supplier No. 62174100MGF
Featured Industries: Food and Beverages

Specifications

CAS 3132-99-8
Flash Point 110°C - closed cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula C18H30O2
MDL Number MFCD00065718
Synonym cis,cis,cis-6,9,12-Octadecatrienoic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 278.43
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less