missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linoleic acid, tech. 75%
CAS: 60-33-3 | C18H32O2 | 280.452 g/mol
$99.95 - $961.01
Chemical Identifiers
| CAS | 60-33-3 |
|---|---|
| Molecular Formula | C18H32O2 |
| Molecular Weight (g/mol) | 280.452 |
| MDL Number | MFCD00064241 |
| InChI Key | OYHQOLUKZRVURQ-HZJYTTRNSA-N |
| Synonym | linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid |
| PubChem CID | 5280450 |
| ChEBI | CHEBI:17351 |
| IUPAC Name | (9Z,12Z)-octadeca-9,12-dienoic acid |
| SMILES | CCCCCC=CCC=CCCCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1526509
|
N/A | 10 g |
Each for $99.95
|
|
|||||
|
AAA1526518
|
N/A | 50 g |
Each for $289.00
|
|
|||||
|
AAA1526530
|
N/A | 250 g |
Each for $961.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C18H32O2 | |
| MFCD00064241 | |
| linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
| CHEBI:17351 | |
| CCCCCC=CCC=CCCCCCCCC(=O)O |
Specifications
| 60-33-3 | |
| 230°C (16 mm) | |
| 75% | |
| 1.4699 | |
| MFCD00064241 | |
| Air sensitive | |
| linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
| CCCCCC=CCC=CCCCCCCCC(=O)O | |
| 280.452 | |
| CHEBI:17351 | |
| 75% | |
| Liquid |
| 0.902 | |
| >110°C (230°F) | |
| C18H32O2 | |
| CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H | |
| 1727101 | |
| 14,5505 | |
| OYHQOLUKZRVURQ-HZJYTTRNSA-N | |
| (9Z,12Z)-octadeca-9,12-dienoic acid | |
| 5280450 | |
| 280.45 | |
| Technical | |
| Linoleic Acid |
Safety and Handling
EINECSNumber : 200-470-9
RTECSNumber : RF9990000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only