missing translation for 'onlineSavingsMsg'
Learn More

Linoleic acid, tech. 75%

Catalog No. AAA1526530
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1526530 250 g
AAA1526509 10 g
AAA1526518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1526530 Supplier Thermo Scientific Chemicals Supplier No. A1526530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60-33-3 | C18H32O2 | 280.452 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60-33-3
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.452
MDL Number MFCD00064241
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
Synonym linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
PubChem CID 5280450
ChEBI CHEBI:17351
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCCC=CCC=CCCCCCCCC(=O)O

Specifications

Density 0.902
Boiling Point 230°C (16 mmHg)
Flash Point >110°C (230°F)
Odor Weak, Characteristic
Assay Percent Range 75%
Refractive Index 1.4699
Linear Formula CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
Quantity 250 g
Beilstein 1727101
Sensitivity Air sensitive
Merck Index 14,5505
Formula Weight 280.45
Percent Purity 75%
Grade Technical
Physical Form Liquid
Chemical Name or Material Linoleic acid
Show More Show Less
EINECSNumber 200-470-9
RTECSNumber RF9990000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only