missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linoleic Acid 85.0+%, TCI America™
$89.23 - $768.93
Chemical Identifiers
| CAS | 60-33-3 |
|---|---|
| Molecular Formula | C18H32O2 |
| Molecular Weight (g/mol) | 280.452 |
| MDL Number | MFCD00064241 |
| InChI Key | OYHQOLUKZRVURQ-HZJYTTRNSA-N |
| Synonym | linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid |
| PubChem CID | 5280450 |
| ChEBI | CHEBI:17351 |
| IUPAC Name | (9Z,12Z)-octadeca-9,12-dienoic acid |
| SMILES | CCCCCC=CCC=CCCCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
L005325ML
|
N/A | 25 mL |
Each for $89.23
|
|
|||||
|
L0053100ML
|
N/A | 100 mL |
Each for $258.21
|
|
|||||
|
L0053500ML
|
N/A | 500 mL |
Each for $768.93
|
|
|||||
Chemical Identifiers
| C18H32O2 | |
| MFCD00064241 | |
| linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
| CHEBI:17351 | |
| CCCCCC=CCC=CCCCCCCCC(=O)O |
Specifications
| 60-33-3 | |
| Yellow | |
| C18H32O2 | |
| linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
| CCCCCC=CCC=CCCCCCCCC(=O)O | |
| 280.452 | |
| CHEBI:17351 | |
| ≥85.0% (GC) | |
| Linoleic Acid |
| -5°C | |
| 230°C | |
| MFCD00064241 | |
| OYHQOLUKZRVURQ-HZJYTTRNSA-N | |
| (9Z,12Z)-octadeca-9,12-dienoic acid | |
| 5280450 | |
| 280.45 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0609
RTECSNumber : RF9990000
TSCA : Yes
Recommended Storage : Refrigerator