missing translation for 'onlineSavingsMsg'
Learn More

Linoleic Acid 85.0+%, TCI America™

Catalog No. L005325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L005325ML 25 mL
L0053100ML 100 mL
L0053500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L005325ML Supplier TCI America Supplier No. L005325ML

Chemical Identifiers

CAS 60-33-3
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.452
MDL Number MFCD00064241
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
Synonym linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
PubChem CID 5280450
ChEBI CHEBI:17351
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCCC=CCC=CCCCCCCCC(=O)O

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 230°C
Quantity 25 mL
Formula Weight 280.45
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Linoleic Acid
EINECSNumber (2)-0609
RTECSNumber RF9990000
TSCA Yes
Recommended Storage Refrigerator