missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LINGO1 (Human) Recombinant Protein
Human LINGO1 full-length ORF ( AAH68558.1, 1 a.a. - 614 a.a.) recombinant protein with GST-tag at N-terminal.
Supplier: Abnova™ H00084894P012
Description
- Sequence: MLAGGVRSMPSPLLACWQPILLLVLGSVLSGSATGCPPRYECSAQDRAVLCHRKRFVAVPEGIPTETRLLDLGKNRIKTLNQDEFASFPHLEELELNENIVSAVEPGAFNNLFNLRTLGLRSNRLKLIPLGVFTGLSNLTKLDISENKIVILLDYMFQDLYNLRSLEVGDNDLVYISHRAFSGLNSLEQLTLEKCNLTSIPTEALSHLHGLIVLRLRHLNINAIRDYSFKRLYRLKVLEISHWPYLDTMTPNCLYGLNLTSLSITHCNLTAVPYLAVRHLVYLRFLNLSYNRISTIEGSMLHELLRLQEIQLVGGQLAVVEPYAFRGLNYLRVLNVSGNQLTTLEESVFHSVGNLETLILDSNPLACDCRLLWVFRRRWRLNFNRQQPTCATPEFVQGKEFKDFPDVLLPNYFTCRRARIRDRKAQQVFVDEGHTVQFVCRADGDPPPAILWLSPRKHLVSAKSNGRLTVFPDGTLEVRYAQVQDNGTYLCIAANAGGNDSMPAHLHVRSYSPDWPHQPNKTFAFISNQPGEGEANSTRATVPFPFDIKTLIIATTMGFISFLGVVLFCLVLLFLWSRGKGNTKHNIEIEYVPRKSDAGISSADAPRKFNMKMI
Specifications
| AAH68558.1 | |
| leucine rich repeat and Ig domain containing 1 | |
| 125% SDS-PAGE Stained with Coomassie Blue | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| LINGO1 | |
| GST |
| 84894 | |
| Wheat germ expression system | |
| 2 μg | |
| FLJ14594, LERN1, LRRN6A, MGC17422, UNQ201 | |
| Wheat Germ (in vitro) | |
| 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |