Learn More
Linezolid, 98%
Linezolid, Zyvox, CAS # 165800-03-3 is a synthetic oxazolidinone antibiotic. | CAS: 165800-03-3 | C16H20FN3O4 | 337.35 g/mol
Supplier: Thermo Scientific Chemicals 460590010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Linezolid is an organoflourine compound and an acetamide
• Linezolid acts by inhibiting initiation complex formation by binding to the 50S ribosomal subunit
Applications
• Linezolid is effective against staphylococcal and enterococcal microbes
• It is most effective against gram-positive organisms, particularly methicillin, vancomycin and penicillin resistant ones
• Suitable for research on antibiotic resistance, cross-reactivity and drug mechanisms
Specifications
| Linezolid | |
| 181.0°C to 183.0°C | |
| Authentic | |
| Glass Bottle | |
| 1 g | |
| Solubility in water: insoluble. | |
| CC(=O)NCC1CN(C(=O)O1)C2=CC(=C(C=C2)N3CCOCC3)F | |
| 337.35 | |
| CHEBI:63607 | |
| 98% |
| 165800-03-3 | |
| 0.5% max. | |
| 97.5% min. (HPLC) | |
| C16H20FN3O4 | |
| linezolid, zyvox, zyvoxid, s-n-3-3-fluoro-4-morpholinophenyl-2-oxooxazolidin-5-yl methyl acetamide, zyvoxam, linezolide, linezlid, zyvoxa, linezolid usan:inn, unii-isq9i6j12j | |
| TYZROVQLWOKYKF-ZDUSSCGKSA-N | |
| N-[[(5S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide | |
| 441401 | |
| 337.35 |
Chemical Identifiers
| 165800-03-3 | |
| 337.35 | |
| linezolid, zyvox, zyvoxid, s-n-3-3-fluoro-4-morpholinophenyl-2-oxooxazolidin-5-yl methyl acetamide, zyvoxam, linezolide, linezlid, zyvoxa, linezolid usan:inn, unii-isq9i6j12j | |
| CHEBI:63607 | |
| CC(=O)NCC1CN(C(=O)O1)C2=CC(=C(C=C2)N3CCOCC3)F |
| C16H20FN3O4 | |
| TYZROVQLWOKYKF-ZDUSSCGKSA-N | |
| 441401 | |
| N-[[(5S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide |
Safety and Handling
GHS H Statement Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Get medical attention/advice if you feel unwell.
GHS Signal Word: Warning
RUO – Research Use Only