missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Linear Barcoded Tubes

Simplify cryotube tracking with durable linear barcodes printed directly on the side of the tube.

Supplier:  Thermo Scientific™ 373530

 View more versions of this product

Catalog No. 12566002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Simplify sample inventory. Thermo Scientific™ Linear Barcoded Tubes feature Thermo Scientific™ Nunc™ and Thermo Scientific™ Nalgene™ Cryotubes with directly printed barcodes that are more robust than labels. Choose from available noncustom options or call for customized options in a wide range of choices of code type, length and format.

Features:

  • Black barcode directly printed onto white background for high contrast and readability even in high-frost conditions
  • Human readable code below the barcode for easy visual identification without a barcode reader
  • Barcodes resistant to IPA, DMSO, 5% bleach, 10% acetic acid and 10% NaOH
  • Noncytotoxic raw materials for added sample security and tubes certified free of DNase and RNase for genomic applications
  • External or internally threading options available
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
Specifications

Chemical Identifiers

Polypropylene
Linear Barcode
Yes
Yes
Yes
Screw Cap

Specifications

12.5 mm
Not Autoclavable
Round
Internal
1800
Yes
Nunc
Yes
0.06086 oz.
Screw Cap
Nunc Barcoded (inkjet interleave 2 of 5) CryoTube Vial
1.8 mL
Polypropylene
48. mm
50
Linear Barcode
Non-sterile
50 Pk., 1800 Cases
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions