missing translation for 'onlineSavingsMsg'
Learn More

Linear Alkylbenzene Sulfonate, Certified, 1000ppm (1mL = 1mg), LabChem™

Catalog No. LC161507
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC161507 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC161507 Supplier LabChem Supplier No. LC161507
Only null left

Specifications

Chemical Name or Material Linear Alkylbenzene Sulfonate
Color Colorless
CAS 7732-18-5, 7664-93-9, 25155-30-0
CAS Max % 99.39, 0.5, 0.11
Linear Formula C18H29NaO3S
Concentration 1000ppm (1mL = 1mg)
Density 1g/mL
Synonym LAS Solution
Solubility Information Soluble in water
Formula Weight 348.48
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Quantity 125 mL
Physical Form Liquid
Show More Show Less