missing translation for 'onlineSavingsMsg'
Learn More

Lindane Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017860
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017860 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017860 Supplier Merck Emd Millipore Supplier No. PHR1244500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Lindane is an organochlorine chemical, widely used as an agricultural insecticide and it also finds applications for the pharmaceutical treatment of infestations of lice and scabies.

Specifications

CAS 58-89-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C6H6 Cl6
MDL Number MFCD00135947
Melting Point 113°C to 115°C
Synonym gamma-hexachlorocyclohexane; gamma-BHC; 1 alpha,2 alpha,3 beta,4 alpha,5 alpha,6 beta-Hexachlorocyclohexane
Type Certified Reference Material
Quantity 500 mg
Formula Weight 290.83
Grade Certified Reference Material
Show More Show Less