missing translation for 'onlineSavingsMsg'
Learn More

Lincomycin Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015339
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015339 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015339 Supplier Merck Emd Millipore Supplier No. PHR16571G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 7179-49-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C18H34N2O6 S · HCl · H2O
MDL Number MFCD00167245
Synonym Lincomycin hydrochloride monohydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 461.01
Grade Certified Reference Material