missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Linavonkibart Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. MA558787
Change view
Click to view available options
Quantity:
100 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA558787 100 μg
MA558788 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA558787 Supplier Invitrogen™ Supplier No. MA558787
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Human Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Linavonkibart, also known as SRK-181, is a fully human IgG4 monoclonal antibody designed to selectively inhibit the latent transforming growth factor-beta isoform 1 (TGFB1). Transforming growth factor-B1 (TGF-B1) is a key member of the TGF-beta family, a group of evolutionarily conserved polypeptide growth factors. The gene encoding TGF-B1 is located on chromosome 19 at the 19q13.2 locus. Structurally, TGF-B1 is synthesized as a precursor, which is then cleaved to produce a mature polypeptide that forms a dimeric protein. Functionally, TGF-B1 plays a crucial role in cellular processes such as proliferation, differentiation, and apoptosis, and is essential for regulating immune responses, maintaining tissue homeostasis, and orchestrating wound healing. Dysregulation of TGF-B1 signaling has been implicated in various pathological conditions including cancer, fibrosis, and autoimmune disorders. The activation of latent TGF-B1 stored in the extracellular matrix is mediated by interaction with integrins and other co-factors that induce conformational changes necessary for its biological activity.

Specifications

Antigen Linavonkibart
Applications ELISA, Functional Assay
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Host Species Human
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG4SP
Show More Show Less