missing translation for 'onlineSavingsMsg'
Learn More

Linalyl Acetate 95.0+%, TCI America™

Catalog No. p-7131794
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L004925ML 25 mL
L0049500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L004925ML Supplier TCI America Supplier No. L004925ML

Chemical Identifiers

CAS 115-95-7
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00008907
InChI Key UWKAYLJWKGQEPM-UHFFFAOYSA-N
Synonym linalyl acetate, linalool acetate, bergamiol, bergamol, linalol acetate, lynalyl acetate, licareol acetate, bergamot mint oil, acetic acid linalool ester, 3,7-dimethyl-1,6-octadien-3-ol acetate
PubChem CID 8294
ChEBI CHEBI:78333
IUPAC Name 3,7-dimethylocta-1,6-dien-3-yl acetate
SMILES CC(=CCCC(C)(C=C)OC(=O)C)C

Specifications

Color Colorless
Boiling Point 220°C
Quantity 25 mL
Formula Weight 196.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Linalyl Acetate
EINECSNumber (2)-2536
RTECSNumber RG5910000
TSCA Yes