missing translation for 'onlineSavingsMsg'
Learn More

Linalyl acetate, 95%, synthetic

Catalog No. AC232300050
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232300050 5 mL Glass bottle
AC232301000 100 mL Glass bottle
AC232305000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC232300050 Supplier Thermo Scientific Chemicals Supplier No. 232300050

CAS: 115-95-7 | C12H20O2 | 196.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115-95-7
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.28
MDL Number MFCD00008907
InChI Key UWKAYLJWKGQEPM-UHFFFAOYSA-N
Synonym linalyl acetate, linalool acetate, bergamiol, bergamol, linalol acetate, lynalyl acetate, licareol acetate, bergamot mint oil, acetic acid linalool ester, 3,7-dimethyl-1,6-octadien-3-ol acetate
PubChem CID 8294
ChEBI CHEBI:78333
IUPAC Name 3,7-dimethylocta-1,6-dien-3-yl acetate
SMILES CC(=CCCC(C)(C=C)OC(=O)C)C

Specifications

Color Colorless
Density 0.9010g/mL
Boiling Point 220°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.452
Linear Formula (CH3)2=CHCH2CH2C(CH3)(OOCCH3)CH=CH2
Quantity 5 mL
Specific Gravity 0.901
Merck Index 15, 5551
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol,alcohol,ether,diethyl phthal,mineral oil,fixed oils
Formula Weight 196.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Linalyl acetate, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 204-116-4
RTECSNumber RG5910000
TSCA TSCA

RUO – Research Use Only