missing translation for 'onlineSavingsMsg'
Learn More

Linalool, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018826
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018826 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018826 Supplier Merck Emd Millipore Supplier No. 517821ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certan Vial

Specifications

CAS 78-70-6
Vapor Pressure 0.17 mmHg (25°C)
Density 0.87 g/mL (at 25°C)
Physical Form Neat
Linear Formula (CH3)2 C=CHCH2CH2C(CH3)(OH)CH=CH2
MDL Number MFCD00008906
Boiling Point 194°C to 197°C (720 mmHg)
Synonym (+/-)-3,7-Dimethyl-1,6-octadien-3-ol; (+/-)-3,7-Dimethyl-3-hydroxy-1,6-octadiene
Quantity 1 mL
Formula Weight 154.25
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less