missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Limonin 95.0+%, TCI America™
$81.64 - $480.34
Chemical Identifiers
| CAS | 1180-71-8 |
|---|---|
| Molecular Formula | C26H30O8 |
| Molecular Weight (g/mol) | 470.52 |
| MDL Number | MFCD00075922 |
| InChI Key | KBDSLGBFQAGHBE-MSGMIQHVSA-N |
| Synonym | limonin, dictamnolactone, obaculactone, citrolimonin, evodin, 7,16-dioxo-7,16-dideoxylimondiol, limoni, limonoic acid, di-delta-lactone, limonine, limonoate d-ring-lactone |
| PubChem CID | 179651 |
| ChEBI | CHEBI:16226 |
| IUPAC Name | (1R,2R,7S,10R,13R,14R,16S,19S,20S)-19-(furan-3-yl)-9,9,13,20-tetramethyl-4,8,15,18-tetraoxahexacyclo[11.9.0.0²,â·.0²,¹â°.0¹â´,¹â¶.0¹â´,²â°]docosane-5,12,17-trione |
| SMILES | CC1(C)O[C@H]2CC(=O)OC[C@@]22[C@H]1CC(=O)[C@]1(C)[C@@H]2CC[C@@]2(C)[C@@H](OC(=O)[C@H]3O[C@@]123)C1=COC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
L0258100MG
|
TCI America
L0258100MG |
100 mg |
Each for $81.64
|
|
|||||
|
L02581G
|
TCI America
L02581G |
1 g |
Each for $480.34
|
|
|||||
Chemical Identifiers
| 1180-71-8 | |
| 470.52 | |
| KBDSLGBFQAGHBE-MSGMIQHVSA-N | |
| 179651 | |
| (1R,2R,7S,10R,13R,14R,16S,19S,20S)-19-(furan-3-yl)-9,9,13,20-tetramethyl-4,8,15,18-tetraoxahexacyclo[11.9.0.0²,â·.0²,¹â°.0¹â´,¹â¶.0¹â´,²â°]docosane-5,12,17-trione |
| C26H30O8 | |
| MFCD00075922 | |
| limonin, dictamnolactone, obaculactone, citrolimonin, evodin, 7,16-dioxo-7,16-dideoxylimondiol, limoni, limonoic acid, di-delta-lactone, limonine, limonoate d-ring-lactone | |
| CHEBI:16226 | |
| CC1(C)O[C@H]2CC(=O)OC[C@@]22[C@H]1CC(=O)[C@]1(C)[C@@H]2CC[C@@]2(C)[C@@H](OC(=O)[C@H]3O[C@@]123)C1=COC=C1 |
Specifications
| 1180-71-8 | |
| White | |
| MFCD00075922 | |
| limonin, dictamnolactone, obaculactone, citrolimonin, evodin, 7,16-dioxo-7,16-dideoxylimondiol, limoni, limonoic acid, di-delta-lactone, limonine, limonoate d-ring-lactone | |
| CC1(C)O[C@H]2CC(=O)OC[C@@]22[C@H]1CC(=O)[C@]1(C)[C@@H]2CC[C@@]2(C)[C@@H](OC(=O)[C@H]3O[C@@]123)C1=COC=C1 | |
| 470.52 | |
| CHEBI:16226 | |
| ≥95.0% (HPLC) | |
| Limonin |
| 298°C | |
| C26H30O8 | |
| 100 mg | |
| KBDSLGBFQAGHBE-MSGMIQHVSA-N | |
| (1R,2R,7S,10R,13R,14R,16S,19S,20S)-19-(furan-3-yl)-9,9,13,20-tetramethyl-4,8,15,18-tetraoxahexacyclo[11.9.0.0²,â·.0²,¹â°.0¹â´,¹â¶.0¹â´,²â°]docosane-5,12,17-trione | |
| 179651 | |
| 470.52 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : RR1122000
TSCA : No
Recommended Storage : Refrigerator