missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Limonene 95.0+%, TCI America™
$43.67 - $214.54
Chemical Identifiers
| CAS | 138-86-3 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.24 |
| MDL Number | MFCD00062992 |
| InChI Key | XMGQYMWWDOXHJM-UHFFFAOYNA-N |
| Synonym | dipentene, limonene, cinene, cajeputene, dl-limonene, kautschin, p-mentha-1,8-diene, dipenten, polylimonene, eulimen |
| PubChem CID | 22311 |
| ChEBI | CHEBI:15384 |
| IUPAC Name | 1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene |
| SMILES | CC(=C)C1CCC(C)=CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
L004625ML
|
N/A | 25 mL |
Each for $43.67
|
|
|||||
|
L0046500ML
|
N/A | 500 mL |
Each for $214.54
|
|
|||||
Chemical Identifiers
| C10H16 | |
| MFCD00062992 | |
| dipentene, limonene, cinene, cajeputene, dl-limonene, kautschin, p-mentha-1,8-diene, dipenten, polylimonene, eulimen | |
| CHEBI:15384 | |
| CC(=C)C1CCC(C)=CC1 |
Specifications
| 138-86-3 | |
| Yellow | |
| C10H16 | |
| 2052 | |
| XMGQYMWWDOXHJM-UHFFFAOYNA-N | |
| 1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene | |
| 22311 | |
| 136.24 | |
| Liquid |
| -96°C | |
| 178°C | |
| MFCD00062992 | |
| dipentene, limonene, cinene, cajeputene, dl-limonene, kautschin, p-mentha-1,8-diene, dipenten, polylimonene, eulimen | |
| CC(=C)C1CCC(C)=CC1 | |
| 136.24 | |
| CHEBI:15384 | |
| ≥95.0% (GC) | |
| (+/-)-Limonene |
Safety and Handling
EINECSNumber : (3)-2245&(7)-0988&(8)-0498
RTECSNumber : OS8100000
TSCA : Yes