missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Limonene 95.0+%, TCI America™

Catalog No. L0046500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0046500ML 500 mL
L004625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0046500ML Supplier TCI America Supplier No. L0046500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138-86-3
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00062992
InChI Key XMGQYMWWDOXHJM-UHFFFAOYNA-N
Synonym dipentene, limonene, cinene, cajeputene, dl-limonene, kautschin, p-mentha-1,8-diene, dipenten, polylimonene, eulimen
PubChem CID 22311
ChEBI CHEBI:15384
IUPAC Name 1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
SMILES CC(=C)C1CCC(C)=CC1

Specifications

Melting Point -96°C
Color Yellow
Boiling Point 178°C
Quantity 500 mL
UN Number 2052
Formula Weight 136.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (+/-)-Limonene
EINECSNumber (3)-2245&(7)-0988&(8)-0498
RTECSNumber OS8100000
TSCA Yes