missing translation for 'onlineSavingsMsg'
Learn More

(-)-Limonene 95.0+%, TCI America™

Supplier:  TCI America L0132500ML

 View more versions of this product

Catalog No. L0132500ML


Only null left

Description

Description

Specifications

Specifications

(-)-Limonene
Yellow
C10H16
500 mL
--limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s--
CC1=CCC(CC1)C(=C)C
136.238
CHEBI:15383
≥95.0% (GC)
5989-54-8
64°C
MFCD00001558
2319
XMGQYMWWDOXHJM-SNVBAGLBSA-N
(4S)-1-methyl-4-prop-1-en-2-ylcyclohexene
439250
136.24
Liquid

Chemical Identifiers

5989-54-8
136.238
XMGQYMWWDOXHJM-SNVBAGLBSA-N
439250
(4S)-1-methyl-4-prop-1-en-2-ylcyclohexene
C10H16
MFCD00001558
--limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s--
CHEBI:15383
CC1=CCC(CC1)C(=C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2245

RTECSNumber : OS8350000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions