missing translation for 'onlineSavingsMsg'
Learn More

(-)-Limonene 95.0+%, TCI America™

Catalog No. p-7131832
Change view
Click to view available options
Quantity:
5 mL
25 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01325ML 5 mL
L013225ML 25 mL
L0132500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L01325ML Supplier TCI America Supplier No. L01325ML

Chemical Identifiers

CAS 5989-54-8
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00001558
InChI Key XMGQYMWWDOXHJM-SNVBAGLBSA-N
Synonym --limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s--
PubChem CID 439250
ChEBI CHEBI:15383
IUPAC Name (4S)-1-methyl-4-prop-1-en-2-ylcyclohexene
SMILES CC1=CCC(CC1)C(=C)C

Specifications

Color Yellow
Boiling Point 64°C
Quantity 5 mL
UN Number 2319
Formula Weight 136.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-Limonene
EINECSNumber (3)-2245
RTECSNumber OS8350000
TSCA Yes