missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Limonene 99.0+%, TCI America™
$112.14 - $377.82
Chemical Identifiers
| CAS | 5989-27-5 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.24 |
| MDL Number | MFCD00062991 |
| InChI Key | XMGQYMWWDOXHJM-JTQLQIEISA-N |
| Synonym | d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene |
| PubChem CID | 440917 |
| ChEBI | CHEBI:15382 |
| IUPAC Name | (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene |
| SMILES | CC(=C)[C@@H]1CCC(C)=CC1 |
Chemical Identifiers
| 5989-27-5 | |
| 136.24 | |
| XMGQYMWWDOXHJM-JTQLQIEISA-N | |
| 440917 | |
| (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene |
| C10H16 | |
| MFCD00062991 | |
| d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene | |
| CHEBI:15382 | |
| CC(=C)[C@@H]1CCC(C)=CC1 |
Specifications
| Clear Liquid at 20°C | |
| 176°C | |
| Colorless | |
| C10H16 | |
| 2319 | |
| XMGQYMWWDOXHJM-JTQLQIEISA-N | |
| (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene | |
| 440917 | |
| 136.24 | |
| (+)-Limonene |
| -75°C | |
| 5989-27-5 | |
| 5 mL | |
| MFCD00062991 | |
| d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene | |
| CC(=C)[C@@H]1CCC(C)=CC1 | |
| 136.24 | |
| CHEBI:15382 | |
| ≥99.0% (GC) |
Safety and Handling
EINECSNumber : (3)-2245
RTECSNumber : GW6360000
TSCA : Yes