missing translation for 'onlineSavingsMsg'
Learn More

(+)-Limonene 99.0+%, TCI America™

Supplier:  TCI America L010525ML

 View more versions of this product

Catalog No. L010525ML


Only null left

Description

Description

Specifications

Specifications

Clear Liquid at 20°C
-75°C
5989-27-5
25 mL
MFCD00062991
d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
CC(=C)[C@@H]1CCC(C)=CC1
136.24
CHEBI:15382
≥99.0% (GC)
(+)-Limonene
176°C
Colorless
C10H16
2319
XMGQYMWWDOXHJM-JTQLQIEISA-N
(4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
440917
136.24

Chemical Identifiers

5989-27-5
136.24
XMGQYMWWDOXHJM-JTQLQIEISA-N
440917
(4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
C10H16
MFCD00062991
d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
CHEBI:15382
CC(=C)[C@@H]1CCC(C)=CC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2245

RTECSNumber : GW6360000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions