missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Limitin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150082
Description
Limitin Polyclonal Antibody for Western Blot, IHC, ICC/IF
Has antiviral and antiproliferative activities. Produced by macrophages and stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.Chemical Identifiers
| Limitin | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q61716 | |
| 15964 | |
| Recombinant Interferon Alpha 11 (IFNa11) (Cys24-Ala190). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Ifna11 | |
| 6030405N23Rik; BGIF; IFN-[a]11; Ifna11; IFN-alpha-11; Ifnz; IFN-z; interferon alpha 11; interferon alpha family, gene 11; interferon alpha-11; interferon zeta; Limitin; Lmtn | |
| Ifna11 | |
| Primary | |
| Protein A |
Specifications
| Limitin | |
| Polyclonal | |
| Unconjugated | |
| Ifna11 | |
| 6030405N23Rik; BGIF; IFN-[a]11; Ifna11; IFN-alpha-11; Ifnz; IFN-z; interferon alpha 11; interferon alpha family, gene 11; interferon alpha-11; interferon zeta; Limitin; Lmtn | |
| Rabbit | |
| Protein A | |
| RUO | |
| 15964 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q61716 | |
| Ifna11 | |
| Recombinant Interferon Alpha 11 (IFNa11) (Cys24-Ala190). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody |