missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Limited Volume Vial Insert

Inserts are ideal for limited and micro volume applications. Options are available for both standard and wide opening 12x32 autosampler type vials.

Supplier:  DWK Life Sciences 60855531

 View more versions of this product

Catalog No. 11417237


Only null left

Description

Description

  • Clear borosilicate glass
  • 60850 series has a conical point design, with polymer springs attached - residual volume 1 mL
  • 60855 series has a flat bottom design - residual volume 25 μL
Specifications

Chemical Identifiers

limited and micro volume applications
100 Pk.

Specifications

200 μL
Flat Bottom
Autosampler Vials 12x32 8 mm Standard Opening
limited and micro volume applications
100 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions