missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LILRA1 Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 53 kDa. Purity is >95% by SDS-PAGE.
LILRA3 is a member of leukocyte immunoglobulin-like receptor gene family. Leukocyte Ig-like receptors (LIRs) are a family of inhibitory and stimulatory immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. LILRA3 lacks a transmembrane region and may act as a soluble receptor for class I MHC antigens. It is expressed in B-cells, and at lower levels in natural killer (NK) cells.
Specifications
Specifications
| Antigen | LILRA1 |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | LILRA1 |
| Gene Accession No. | O75019 |
| Gene Alias | CD85 antigen-like family member I; CD85I; leucocyte Ig-like receptor A1; leukocyte immunoglobulin like receptor A1; leukocyte immunoglobulin-like receptor 6; Leukocyte immunoglobulin-like receptor subfamily A member 1; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1; LILRA1; LIR6; LIR-6 |
| Gene Symbols | LILRA1 |
| Show More |