Learn More
BIOTIX™ xPIPETTE Adjustable Single Channel Pipette
Patented ejection technology reduces fatigue and your risk of repetitive stress injuries while providing superior accuracy and precision.
Supplier:  BIOTIX™ 30459891
 View more versions of this product
Catalog No. 12-111-394
Get Extra Biotix Pipette Products with Purchase
View all promotions
Only null left
Description
Description
  • Compatible with Biotix xTIP4™ and Rainin™ LTS™ pipette tips
  • Superior accuracy and precision with better ergonomics
  • Tip cone features a positive stop that works with Biotix xTIP4 tips to create identical tip alignment and sealing on every tip
  • Up to 5X less ejection force than traditional pipetting systems
  • Low drag seals and light spring forces produce a smooth draw and increased control
  • Easy to reach ejector button is cushioned to reduce thumb impact and stress when ejecting tips
  • Volume lock ensures accurate and precise measurements without volume drift
  • Superior accuracy and precision for more reliable data
  • Certifications
    ISO 9001
    Specifications
    Specifications
    xTIP4 and LTS- 20, 10
    Red
    xTIP 4 Pipette Tip
    PVDF
    1
    No
    Adjustable Single Channel Pipette
    xTIP4 pipette tips
    6 cm
    24 cm
    Autoclavable
    Single-use
    Ergonomic
    Biotix xPIPETTE
    xPIPETTE
    Non-sterile
    0.5 to 10 μL
    Calibration Certificate
    20 mm
    Product Suggestions
    Product Suggestions