missing translation for 'onlineSavingsMsg'
Learn More

Light's Solution, Oxidation-Reduction Potential (ORP) Standard, Ricca Chemical

Catalog No. 433032
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
433032 1 L
433016 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 433032 Supplier Ricca Chemical Company Supplier No. 433032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Light's Solution
Packaging Natural Poly Bottle
Name Note Oxidation-Reduction Potential (ORP) Standard
CAS Min % 82.0, 8.51, 3.97, 3.23
CAS Max % 86.0, 8.86, 4.13, 3.36
CAS 7664-93-9, 7783-83-7, 7783-85-9
Physical Form Liquid
Color Yellow
Melting Point Approximately 0°C
Quantity 1 L