missing translation for 'onlineSavingsMsg'
Learn More

Light's Solution Oxidation-Reduction Potential (ORP) Standard, Reagents

Catalog No. p-200021494
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334851 1 L
01334850 4 L
01334849 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334851 Supplier Labchem Inc Supplier No. CL0003001A
Only null left

Light's Solution Oxidation-Reduction Potential (ORP) Standard, Reagent, use for Redox. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Light's Solution Oxidation-Reduction Potential (ORP) Standard
Packaging Poly Bottle
CAS 7732-18-5, 7783-85-9, 7664-93-9
Specific Gravity 1.0857
Physical Form Liquid
Color Yellow
pH <1
Boiling Point 100°C
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Show More Show Less