missing translation for 'onlineSavingsMsg'
Learn More

Light Mineral Oil, NF, Spectrum™ Chemical

Catalog No. p-7157818
Change view
Click to view available options
Quantity:
1 L
20 L
200 L
4 L
4 x 4 L
500 mL
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Catalog No. Quantity Packaging
18603883 500 mL Amber Glass Bottle
18603884 1 L Amber Glass Bottle
18603885 4 L Amber Glass Bottle
18603886 4 x 4 L Amber Glass Bottle
18603887 20 L Poly Drum
18603888 200 L Drum
6 options

Catalog No. 18603883

Supplier: Spectrum Chemical Mfg Cor MI501500MLGL

Only null left

MI501, 8042-47-5

Spectrum™ Chemical Light Mineral Oil, NF is used as a vehicle and humectant in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8042-47-5

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 mL
Grade NF