missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Light Green SF Yellowish

CAS: 5141-20-8 | C37H34N2Na2O9S3 | 792.844 g/mol

Supplier:  Thermo Scientific Chemicals B2333009

 View more versions of this product

Catalog No. AAB2333009


Only null left

Description

Description

Useful as a general plasma stain, as a histological stain, and a cytological counterstain

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Light Green SF Yellowish
5141-20-8
MFCD00012121
14,5485
DGOBMKYRQHEFGQ-UHFFFAOYSA-L
disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
21223
792.86
∼288°C (decomposition)
C37H34N2Na2O9S3
5717828
Acid Green 5; C.I. 42095
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]
792.844
CHEBI:87065
10 g

Chemical Identifiers

5141-20-8
792.844
DGOBMKYRQHEFGQ-UHFFFAOYSA-L
21223
disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
C37H34N2Na2O9S3
MFCD00012121
Acid Green 5; C.I. 42095
CHEBI:87065
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 225-906-5

RTECSNumber : BQ4900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only