missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Light Green SF Yellowish

Catalog No. p-7026962
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2333009 10 g
AAB2333018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2333009 Supplier Thermo Scientific Chemicals Supplier No. B2333009
Only null left

CAS: 5141-20-8 | C37H34N2Na2O9S3 | 792.844 g/mol

Useful as a general plasma stain, as a histological stain, and a cytological counterstain

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5141-20-8
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.844
MDL Number MFCD00012121
InChI Key DGOBMKYRQHEFGQ-UHFFFAOYSA-L
Synonym Acid Green 5; C.I. 42095
PubChem CID 21223
ChEBI CHEBI:87065
IUPAC Name disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point ∼288°C (decomposition)
Beilstein 5717828
Merck Index 14,5485
Formula Weight 792.86
Quantity 10 g
Chemical Name or Material Light Green SF Yellowish
Hazard Category Warning
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 225-906-5
RTECSNumber BQ4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only