Learn More
Invitrogen™ ReadyProbes™ Cell Viability Imaging Kit, Blue/Green
Description
Includes
3 x 2.5mL NucBlue Live, 3 x 2.5mL NucGreen Dead
ReadyProbes™ Cell Viability Imaging Kit, Blue/Green contains NucBlue™ Live reagent (Hoechst 33342) and NucGreen™ Dead reagent. Just add 2 drops of each stable reagent (at room temperature) to 1mL of cell growth media, then determine viability by counting total vs dead cells. NucBlue™ Live reagent stains the nuclei of all the cells and can be detected with a standard DAPI filter. NucGreen™ Dead reagent stains only the nuclei of cells with compromised plasma membrane integrity and is detected using a standard FITC/GFP (green) filter set.
- NucBlue™ Live reagent: stains the nuclei of all cells; detected with a standard DAPI filter (excitation/emission maxima: 360/460nm)
- NucGreen™ Dead reagent: stains only the nuclei of dead cells with compromised plasma membranes; detected with standard FITC/GFP (green) filter set (excitation/emission maxima: 504/523nm)
Suggestions for use
- NucBlue™ Live and NucGreen™ Dead reagents may be added directly to cells in full growth media or a compatible buffer solution.
- In most cases, 2 drops/mL and an incubation time of 5 to 30 minutes will give bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained.
Specifications
Specifications
| Cell Type | Mammalian Cells, Eukaryotic Cells |
| Color | Blue, Green |
| Content And Storage | Store at 2° to 8°C, or at room temperature. |
| Description | ReadyProbes Cell Viability Imaging Kit, Blue/Green |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Cell Viability Imaging Kit |
| Dye Type | 3 x 2.5 mL NucBlue™ Live |
| Format | Dropper bottles |
| Show More |
Frequently Asked Questions (FAQs)
This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.
We have validated the following kits for use on the Countess II FL Automated Cell Counter:
LIVE/DEAD Viabilty/Cytoxicity Kit (Cat. No. L3224) containing calcein AM and ethidium homodimer-1
ReadyProbes Cell Viability Imaging Kit, Blue/Green (Cat. No. R37609)
ReadyProbes Cell Viability Imaging Kit, Blue/Red (Cat. No. R37610) containing NucBlue Live/NucGreen Dead and NucBlue Live/propidium iodide
See this Application Note for details - https://www.thermofisher.com/content/dam/LifeTech/global/life-sciences/Laboratory%20Instruments/Files/0415/CO014723-Countess-II-FL-Viability-Appnote_FHR.pdf.
The NucBlue Live reagent stains the nuclei of all cells, while NucGreen Dead reagent stains only the nuclei of dead cells.
For Research Use Only. Not for use in diagnostic procedures.