missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017766
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017766 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017766 Supplier Merck Emd Millipore Supplier No. PHR1669100MG
Only null left

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Pharmaceutical (small molecule)

Specifications

CAS 87-62-7
Vapor Pressure <0.01 mmHg (20°C)
Density 0.984 g/mL (at 25°C)
Flash Point 91°C - Closed Cup
Linear Formula (CH3)2C6H3 NH2
MDL Number MFCD00007747
Melting Point 10°C to 12°C
Boiling Point 214°C (739 mmHg)
Synonym 2,6-Dimethylaniline; 2,6-Dimethylaniline; 2,6-Xylidine; 2-Amino-1,3-dimethylbenzene; 2-Amino-m-xylene
Type Certified Reference Material
Quantity 100 mg
Formula Weight 121.18
Grade Certified Reference Material
Show More Show Less